Phenol, 4-[(1S)-1-aminoethyl]- 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-4-(1-Aminoethyl)phenol
H63269.03
H63269.06
H63269.03EA
218
EUR
H63269.03
H63269.06
Phenol, 4-[(1S)-1-aminoethyl]- 97%
Phenol, 4-[(1S)-1-aminoethyl]-
Purity: 97%
CAS No.: 221670-72-0
Molecular Formula: C8H11NO
Formula Weight: 137.18
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
UN No.: UN3259
MDL No.: MFCD03844646
Formel:
C₈H₁₁NO |
MDL:
MFCD03844646 CAS-Nummer: 221670-72-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...