Fmoc-Met(O)-OH 98%
Lieferant: Thermo Scientific
Synonyme:
N-Fmoc-L-methionine sulphoxide
H63271.03
H63271.06
H63271.03EA
39.9
EUR
H63271.03
H63271.06
Fmoc-Met(O)-OH 98%
Fmoc-Met(O)-OH
Purity: 98%
CAS No.: 76265-70-8
Molecular Formula: C20H21NO5S
Formula Weight: 387.45
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00077064
Formel:
C₂₀H₂₁NO₅S Molecular Weight: 387.45 g/mol |
MDL:
MFCD00077064 CAS-Nummer: 76265-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...