(S)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate 98%

Lieferant: Thermo Scientific

Synonyme: Ngamma-Trityl-L-asparagine hydrate, H-Asn(Trt)-OH hydrate, H-Asn(Trt)-OH·H₂O, Nγ-Trityl-L-Asparagine hydrate

H63285.03 H63285.06
H63285.03EA 35.3 EUR
H63285.03 H63285.06
(S)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate 98%
(S)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate

Purity: 98% CAS No.: 132388-58-0 Molecular Formula: C23H22N2O3.xH2O Formula Weight: 392.45(anhy) Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00153422

Formel: C₂₃H₂₄N₂O₄
Molecular Weight: 392,44 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00153422
CAS-Nummer: 132388-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR