3-(2-Thienyl)-L-alanine 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: H-β-(2-Thienyl)-Ala-OH, 3-(2-Thienyl)-L-alanine, (S)-2-Amino-3-(thiophen-2-yl)propanoic acid

H63316.03 H63316.06
H63316.03EA 189 EUR
H63316.03 H63316.06
3-(2-Thienyl)-L-alanine 95%
3-(2-Thienyl)-L-alanine

Purity: 95% CAS No.: 22951-96-8 Molecular Formula: C7H9NO2S Formula Weight: 171.21 Physical Form: Solid Appearance: Off-white Melting Point: 255-263° dec Beilstein Reference: 82872 Optical Rotation: -31°(c=1 in water) MDL No.: MFCD00064335

Formel: C₇H₉NO₂S
Molecular Weight: 171,214 g/mol
Schmelzpunkt: 255…263 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064335
CAS-Nummer: 22951-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR