4-Fluor-3-nitrotoluol 98%

Lieferant: Thermo Fisher Scientific
Achtung

H63317.14 H63317.22
H63317.14EA 72.8 EUR
H63317.14 H63317.22
4-Fluor-3-nitrotoluol 98%
4-Fluor-3-nitrotoluol

Purity: 98% CAS No.: 446-11-7 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Physical Form: Liquid Appearance: Red-brown Boiling Point: 241° dec Beilstein Reference: 2253853 UN No.: UN2810 MDL No.: MFCD00007060

Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
Siedepunkt: 241 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007060
CAS-Nummer: 446-11-7
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR