(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
D-Phenylalanine tert-butyl ester hydrochloride, H-D-Phe-OtBu·HCl, H-D-Phe-OtBu.HCl
H63325.03
H63325.06
H63325.03EA
30.1
EUR
H63325.03
H63325.06
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 98%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride
Purity: 98%
CAS No.: 3403-25-6
Molecular Formula: C13H19NO2.HCl
Formula Weight: 257.76
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00153455
Formel:
C₁₃H₁₉NO₂·HCl Molecular Weight: 257.8 g/mol |
MDL:
MFCD00153455 CAS-Nummer: 3403-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...