Fmoc-Orn(Alloc)-OH 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-Orn(Aloc)-OH, Nδ-Allyloxycarbonyl-Nα-Fmoc-L-ornithine
H63331.06
H63331.14
H63331.06EA
109
EUR
H63331.06
H63331.14
Fmoc-Orn(Alloc)-OH 95%
Fmoc-Orn(Alloc)-OH
Purity: 95%
CAS No.: 147290-11-7
Molecular Formula: C24H26N2O6
Formula Weight: 438.47
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00798637
Formel:
C₂₄H₂₆N₂O₆ Molecular Weight: 438.48 g/mol |
MDL:
MFCD00798637 CAS-Nummer: 147290-11-7 |