2,4-Dichloro-3-cyanopyridine 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,4-Dichloropyridine-3-carbonitrile, 2,4-Dichloronicotinonitrile
H63347.03
H63347.06
H63347.03EA
91.9
EUR
H63347.03
H63347.06
2,4-Dichloro-3-cyanopyridine 95%
2,4-Dichloro-3-cyanopyridine
Purity: 95%
CAS No.: 180995-12-4
Molecular Formula: C6H2Cl2N2
Formula Weight: 173.00
Physical Form: Solid
UN No.: UN3439
MDL No.: MFCD09750302
Formel:
C₆H₂Cl₂N₂ Molecular Weight: 172,996 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09750302 CAS-Nummer: 180995-12-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...