Fmoc-Glu(OMe)-OH 97%
Lieferant: Thermo Scientific
Synonyme:
N-Fmoc-L-glutamic acid 5-methyl ester
H63358.06
H63358.14
H63358.06EA
53.4
EUR
H63358.06
H63358.14
Fmoc-Glu(OMe)-OH 97%
Fmoc-Glu(OMe)-OH
Purity: 97%
CAS No.: 145038-50-2
Molecular Formula: C21H21NO6
Formula Weight: 383.39
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD01076132
Formel:
C₂₁H₂₁NO₆ |
MDL:
MFCD01076132 CAS-Nummer: 145038-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...