4-Thiomorpholinoaniline 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-(4-Thiomorpholinyl)aniline
H63360.MD
H63360.03
H63360.MDEA
60.4
EUR
H63360.MD
H63360.03
4-Thiomorpholinoaniline 97%
4-Thiomorpholinoaniline
Purity: 97%
CAS No.: 22589-35-1
Molecular Formula: C10H14N2S
Formula Weight: 194.3
Storage Temperature: 2°C to 8°C
Physical Form: Solid
UN No.: UN2811
MDL No.: MFCD01625958
Formel:
C₁₀H₁₄N₂S |
MDL:
MFCD01625958 CAS-Nummer: 22589-35-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...