D-2-Cyclohexylglycine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
(R)-alpha-Aminocyclohexylacetic Acid, H-D-Chg-OH
H63381.03
H63381.06
H63381.03EA
44.1
EUR
H63381.03
H63381.06
D-2-Cyclohexylglycine 95%
D-2-Cyclohexylglycine
Purity: 95%
CAS No.: 14328-52-0
Molecular Formula: C8H15NO2
Formula Weight: 157.21
Physical Form: Solid
Beilstein Reference: 3196806
MDL No.: MFCD01311678
Formel:
C₈H₁₅NO₂ Molecular Weight: 157.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311678 CAS-Nummer: 14328-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...