(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid 95%
Lieferant: Thermo Scientific
Synonyme:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-alanine, Fmoc-N-Me-Ala-OH, N-Fmoc-N-methyl-L-alanine
H63411.06
H63411.14
H63411.06EA
86.5
EUR
H63411.06
H63411.14
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid
Purity: 95%
CAS No.: 84000-07-7
Molecular Formula: C19H19NO4
Formula Weight: 325.36
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 140°
Beilstein Reference: 4329998
Optical Rotation: -18°(c=1 in DMF)
MDL No.: MFCD00153384
Formel:
C₁₉H₁₉NO₄ Molecular Weight: 325,4 g/mol Schmelzpunkt: 140 °C |
MDL:
MFCD00153384 CAS-Nummer: 84000-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...