(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane 97%
Lieferant: Thermo Scientific
Synonyme: (2R,5R)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane
H63420.MC H63420.ME
H63420.MCEA 72.1 EUR
H63420.MC H63420.ME
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane 97%
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Purity: 97% CAS No.: 1044256-04-3 Molecular Formula: C15H21O2P Formula Weight: 264.30 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.105 Refractive Index: 1.5720 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Optical Rotation: -38°(c=1 in chloroform) MDL No.: MFCD09842713
Formel: C₁₅H₂₁O₂P
Molecular Weight: 264,305 g/mol
Dichte: 1,105
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD09842713
CAS-Nummer: 1044256-04-3