Fmoc-N-methyl-L-leucine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Fmoc-N-methyl-L-leucine, Fmoc-N-Me-Leu-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine
H63453.03
H63453.06
H63453.03EA
82.7
EUR
H63453.03
H63453.06
Fmoc-N-methyl-L-leucine 98%
Fmoc-N-methyl-L-leucine
Purity: 98%
CAS No.: 103478-62-2
Molecular Formula: C22H25NO4
Formula Weight: 367.44
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 113-116°
Beilstein Reference: 3630430
Optical Rotation: -20°(c=1 in DMF)
MDL No.: MFCD00151933
Formel:
C₂₂H₂₅NO₄ Molecular Weight: 367.445 g/mol Schmelzpunkt: 113…116 °C |
MDL:
MFCD00151933 CAS-Nummer: 103478-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...