Fmoc-D-allo-isoleucine 98%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-D-allo-Ile-OH, N-Fmoc-D-allo-isoleucine
H63455.03
H63455.06
H63455.03EA
224
EUR
H63455.03
H63455.06
Fmoc-D-allo-isoleucine 98%
Fmoc-D-allo-isoleucine
Purity: 98%
CAS No.: 118904-37-3
Molecular Formula: C21H23NO4
Formula Weight: 353.41
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00077062
Formel:
C₂₁H₂₃NO₄ Molecular Weight: 353,418 g/mol |
MDL:
MFCD00077062 CAS-Nummer: 118904-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...