H-D-Glu(OtBu)-OH 95%

Lieferant: Thermo Scientific

Synonyme: D-Glutamic acid 5-tert-butyl ester

H63463.03 H63463.06 H63463.14
H63463.03EA 35.4 EUR
H63463.03 H63463.06 H63463.14
H-D-Glu(OtBu)-OH 95%
H-D-Glu(OtBu)-OH

Purity: 95% CAS No.: 45125-00-6 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00153437

Formel: C₉H₁₇NO₄
MDL: MFCD00153437
CAS-Nummer: 45125-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR