Boc-Lys(Z)-OSu 95%
Lieferant: Thermo Scientific
Synonyme:
Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine N-succinimidyl ester
H63470.06
H63470.14
H63470.06EA
89
EUR
H63470.06
H63470.14
Boc-Lys(Z)-OSu 95%
Boc-Lys(Z)-OSu
Purity: 95%
CAS No.: 34404-36-9
Molecular Formula: C23H31N3O8
Formula Weight: 477.51
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 108-117°
Beilstein Reference: 1559393
Optical Rotation: -21° (c=2 in dioxane)
MDL No.: MFCD00037915
Formel:
C₂₃H₃₁N₃O₈ Molecular Weight: 477,514 g/mol Schmelzpunkt: 108…117 °C |
MDL:
MFCD00037915 CAS-Nummer: 34404-36-9 EINECS: 251-999-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...