Methyl-2,5-difluor-4-nitrobenzoat 95%

Lieferant: Thermo Scientific

Synonyme: Methyl 2,5-difluoro-4-nitrobenzoate

H63472.03 H63472.06
H63472.03EA 150 EUR
H63472.03 H63472.06
Methyl-2,5-difluor-4-nitrobenzoat 95%
Methyl-2,5-difluor-4-nitrobenzoat

Purity: 95% CAS No.: 924868-81-5 Molecular Formula: C8H5F2NO4 Formula Weight: 217.13 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD08689670

Formel: C₈H₅F₂NO₄
Molecular Weight: 217,13 g/mol
MDL: MFCD08689670
CAS-Nummer: 924868-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR