Methyl thiazole-2-carboxylate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-thiazolecarboxylate

H63496.03 H63496.06
H63496.03EA 88.4 EUR
H63496.03 H63496.06
Methyl thiazole-2-carboxylate 95%
Methyl thiazole-2-carboxylate

Purity: 95% CAS No.: 55842-56-3 Molecular Formula: C5H5NO2S Formula Weight: 143.16 Physical Form: Solid MDL No.: MFCD18075580

Formel: C₅H₅NO₂S
Molecular Weight: 143,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18075580
CAS-Nummer: 55842-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR