H-Thr(tBu)-OMe·HCl 95%

Lieferant: Thermo Scientific

Synonyme: Methyl O-(tert-butyl)-L-threoninate hydrochloride, O-tert-Butyl-L-threonine methyl ester hydrochloride, H-Thr(tBu)-OMe·HCl

H63529.03 H63529.06
H63529.03EA 46.4 EUR
H63529.03 H63529.06
H-Thr(tBu)-OMe·HCl 95%
H-Thr(tBu)-OMe·HCl

Purity: 95% CAS No.: 71989-43-0 Molecular Formula: C9H19NO3.HCl Formula Weight: 225.71 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 4716005 MDL No.: MFCD00077111

Formel: C₉H₂₀ClNO₃
Molecular Weight: 225,713 g/mol
MDL: MFCD00077111
CAS-Nummer: 71989-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR