3-aminopyrazine-2-carboxamide 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Aminopyrazine-2-carboxamide

H63553.03 H63553.06
H63553.03EA 181 EUR
H63553.03 H63553.06
3-aminopyrazine-2-carboxamide 96%
3-aminopyrazine-2-carboxamide

Purity: 96% CAS No.: 32587-10-3 Molecular Formula: C5H6N4O Formula Weight: 138.13 Physical Form: Solid MDL No.: MFCD03407446

Formel: C₅H₆N₄O
Molecular Weight: 138,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407446
CAS-Nummer: 32587-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR