DL-Phenylalaninol 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Amino-3-phenyl-1-propanol, DL-2-Amino-3-phenyl-1-propanol, 2-Amino-3-phenylpropan-1-ol
H63562.03
H63562.06
H63562.14
H63562.03EA
52
EUR
H63562.03
H63562.06
H63562.14
DL-Phenylalaninol 96%
DL-Phenylalaninol
Purity: 96%
CAS No.: 16088-07-6
Molecular Formula: C9H13NO
Formula Weight: 151.21
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 69-73°
Beilstein Reference: 2208237
UN No.: UN3259
MDL No.: MFCD00066689
Formel:
C₉H₁₃NO Molecular Weight: 151,21 g/mol Schmelzpunkt: 69…73 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00066689 CAS-Nummer: 16088-07-6 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...