L-Glutamic acid dibenzyl ester hydrochloride 98%
Lieferant: Thermo Scientific
Synonyme:
H-Glu(OBzl)-OBzl.HCI, H-Glu(OBzl)-OBzl·HCl, L-Glutamic acid dibenzyl ester hydrochloride, Dibenzyl L-Glutamate hydrochloride
H63598.03
H63598.06
H63598.03EA
43.5
EUR
H63598.03
H63598.06
L-Glutamic acid dibenzyl ester hydrochloride 98%
L-Glutamic acid dibenzyl ester hydrochloride
Purity: 98%
CAS No.: 4561-10-8
Molecular Formula: C19H21NO4.HCl
Formula Weight: 363.84
Physical Form: Powder
Appearance: White
MDL No.: MFCD00035103
Formel:
C₁₉H₂₁NO₄·HCl Molecular Weight: 363,838 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035103 CAS-Nummer: 4561-10-8 |