N-Boc-thiourea 97%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: tert-Butyl carbamothioylcarbamate
H63599.03 H63599.06
H63599.03EA 88.6 EUR
H63599.03 H63599.06
N-Boc-thiourea 97%
N-Boc-thiourea
Purity: 97% CAS No.: 268551-65-1 Molecular Formula: C6H12N2O2S Formula Weight: 176 Physical Form: Powder Appearance: Off-white Melting Point: 142° dec MDL No.: MFCD04039987
Formel: C₆H₁₂N₂O₂S
Molecular Weight: 176.234 g/mol
Schmelzpunkt: 142 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD04039987
CAS-Nummer: 268551-65-1