6-Brom-2,4-dichlorchinazolin 97%

Lieferant: Thermo Scientific
Achtung

H63603.03 H63603.06
H63603.03EA 173 EUR
H63603.03 H63603.06
6-Brom-2,4-dichlorchinazolin 97%
6-Bromo-2,4-dichloroquinazoline

Purity: 97% CAS No.: 102393-82-8 Molecular Formula: C8H3BrCl2N2 Formula Weight: 277.93 Physical Form: Solid Appearance: Off-white MDL No.: MFCD09744007

Formel: C₈H₃BrCl₂N₂
Molecular Weight: 277,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09744007
CAS-Nummer: 102393-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR