(R)-2-Amino-3-(tritylthio)propanamide 98%
Lieferant: Thermo Scientific
Synonyme:
H-Cys(Trt)-NH2, H-Cys(Trt)-NH₂, S-Trityl-L-cysteinamide
H63628.03
H63628.06
H63628.03EA
46
EUR
H63628.03
H63628.06
(R)-2-Amino-3-(tritylthio)propanamide 98%
(R)-2-Amino-3-(tritylthio)propanamide
Purity: 98%
CAS No.: 166737-85-5
Molecular Formula: C22H22N2OS
Formula Weight: 362.49
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD22126061
Formel:
C₂₂H₂₂N₂OS Molecular Weight: 362,49 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22126061 CAS-Nummer: 166737-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...