1-Methyl-5-(2-methyl-2-propanyl)-L-glutamathydrochlorid 98%
Lieferant: Thermo Scientific
Synonyme:
L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester Hydrochloride, H-Glu(OtBu)-OMe·HCl, H-Glu(OtBu)-OMe.HCl
H63661.03
H63661.06
H63661.03EA
78
EUR
H63661.03
H63661.06
1-Methyl-5-(2-methyl-2-propanyl)-L-glutamathydrochlorid 98%
1-Methyl-5-(2-methyl-2-propanyl)-L-glutamathydrochlorid
Purity: 98%
CAS No.: 6234-01-1
Molecular Formula: C10H19NO4.HCl
Formula Weight: 253.72
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 3572140
MDL No.: MFCD00153438
Formel:
C₁₀H₁₉NO₄ Molecular Weight: 253,72 g/mol |
MDL:
MFCD00153438 CAS-Nummer: 6234-01-1 EINECS: 228-348-0 |