O-Benzyl-L-tyrosine 98%
Lieferant: Thermo Scientific
Synonyme:
H-Tyr(Bzl)-OH, H-Tyr(Bn)-OH
H63675.06
H63675.14
H63675.06EA
66.3
EUR
H63675.06
H63675.14
O-Benzyl-L-tyrosine 98%
O-Benzyl-L-tyrosine
Purity: 98%
CAS No.: 16652-64-5
Molecular Formula: C16H17NO3
Formula Weight: 271.31
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 259° dec
Beilstein Reference: 2659642
Optical Rotation: -10°(c=1 in acetic acid: water (4:1))
MDL No.: MFCD00002605
Formel:
C₁₆H₁₇NO₃ Molecular Weight: 271,32 g/mol Schmelzpunkt: 259 °C (Zers.) |
MDL:
MFCD00002605 CAS-Nummer: 16652-64-5 EINECS: 240-699-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...