Boc-Abu-OH 95%
Lieferant: Thermo Scientific
Synonyme:
(S)-2-(tert-Butoxycarbonylamino)butyric acid, Boc-L-2-aminobutanoic acid, BOC-L-α-Aminobutyric acid, Boc-L-Abu-OH, (S)-2-(Boc-amino)butyric acid
H63686.03
H63686.06
H63686.14
H63686.03EA
46.4
EUR
H63686.03
H63686.06
H63686.14
Boc-Abu-OH 95%
Boc-Abu-OH
Purity: 95%
CAS No.: 34306-42-8
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Physical Form: Solid
Beilstein Reference: 6801706
Optical Rotation: -18°(c=1 in methanol)
MDL No.: MFCD00037267
Formel:
C₉H₁₇NO₄ Molecular Weight: 203,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037267 CAS-Nummer: 34306-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...