2-Boc-2-azabicyclo[2.2.1]hept-5-ene 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
tert-Butyl 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylate, 2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene, 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylic acid tert-Butyl ester, 2-Boc-2-azabicyclo[2.2.1]hept-5-ene
H63698.03
H63698.06
H63698.03EA
112
EUR
H63698.03
H63698.06
2-Boc-2-azabicyclo[2.2.1]hept-5-ene 98%
2-Boc-2-azabicyclo[2.2.1]hept-5-ene
Purity: 98%
CAS No.: 188345-71-3
Molecular Formula: C11H17NO2
Formula Weight: 195.26
Physical Form: Liquid
Appearance: Yellow
MDL No.: MFCD22416354
Formel:
C₁₁H₁₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22416354 CAS-Nummer: 188345-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...