Acetoin 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Acetylmethylcarbinol, 3-Hydroxybutanon, 3-Hydroxy-2-butanon
A13752.0B
A13752.18
A13752.30
A13752.0BEA
458
EUR
A13752.0B
A13752.18
A13752.30
Acetoin 95%
Acetoin
Purity: 95%
CAS No.: 513-86-0
Molecular Formula: C4H8O2
Formula Weight: 88.11
Storage Temperature: 2°C to 8°C
Flash Point: 41°(106°F)
Odour: Characteristic
Boiling Point: 148°
Beilstein Reference: 385636
Merck Reference: 14,64
UN No.: 2621
MDL No.: MFCD00038696
Formel:
C₄H₈O₂ Molecular Weight: 88,11 g/mol Siedepunkt: 148 °C Schmelzpunkt: 20 °C Dichte: 1,013 Flammpunkt: 41 °C (106 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00038696 CAS-Nummer: 513-86-0 EINECS: 208-174-1 UN: 2621 ADR: 3,III Merck Index: 14,64 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...