(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
Lieferant: Thermo Scientific
Synonyme:
Boc-Cys(Bzl)-OH, N-Boc-S-benzyl-L-cysteine
H63736.06
H63736.14
H63736.06EA
31.6
EUR
H63736.06
H63736.14
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid
Purity: 98%
CAS No.: 5068-28-0
Molecular Formula: C15H21NO4S
Formula Weight: 311.4
Physical Form: Solid
Melting Point: 86-88°
Beilstein Reference: 2000417
Optical Rotation: -44°(c=1 in acetic acid)
MDL No.: MFCD00065567
Formel:
C₁₅H₂₁NO₄S Molecular Weight: 311,4 g/mol Schmelzpunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065567 CAS-Nummer: 5068-28-0 EINECS: 225-772-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...