Fmoc-Lys(Alloc)-OH 95%
Lieferant: Thermo Scientific
Synonyme:
Nepsilon-Allyloxycarbonyl-Nα-Fmoc-L-lysine, Fmoc-Lys(Aloc)-OH
H63768.03
H63768.06
H63768.03EA
51
EUR
H63768.03
H63768.06
Fmoc-Lys(Alloc)-OH 95%
Fmoc-Lys(Alloc)-OH
Purity: 95%
CAS No.: 146982-27-6
Molecular Formula: C25H28N2O6
Formula Weight: 452.50
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 5893081
Optical Rotation: -12°(c=1 in DMF)
MDL No.: MFCD00190872
Formel:
C₂₅H₂₈N₂O₆ Molecular Weight: 452,507 g/mol |
MDL:
MFCD00190872 CAS-Nummer: 146982-27-6 |