Fmoc-aspartic acid-4-methyl ester 97%
Lieferant: Thermo Scientific
Synonyme:
N-Fmoc-L-aspartic acid 4-methyl ester, Fmoc-Asp(OMe)-OH
H63774.03
H63774.06
H63774.03EA
35.4
EUR
H63774.03
H63774.06
Fmoc-aspartic acid-4-methyl ester 97%
Fmoc-aspartic acid-4-methyl ester
Purity: 97%
CAS No.: 145038-53-5
Molecular Formula: C20H19NO6
Formula Weight: 369.37
Physical Form: Solid
MDL No.: MFCD08458573
Formel:
C₂₀H₁₉NO₆ Molecular Weight: 369,4 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458573 CAS-Nummer: 145038-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...