3-Iod-L-tyrosin 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
H63789.03
H63789.06
H63789.03EA
49.9
EUR
H63789.03
H63789.06
3-Iod-L-tyrosin 98%
3-Iod-L-tyrosin
Purity: 98%
CAS No.: 70-78-0
Molecular Formula: C9H10INO3
Formula Weight: 307.09
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 210° dec
Beilstein Reference: 2941266
MDL No.: MFCD00002608
Formel:
C₉H₁₀INO₃ Molecular Weight: 307.1 g/mol Schmelzpunkt: 210 °C (Zers.) |
MDL:
MFCD00002608 CAS-Nummer: 70-78-0 EINECS: 200-744-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...