2-Methyl-6-chinolincarbonsäure 97%

Lieferant: Thermo Scientific
Achtung

H63790.03 H63790.06 H63790.14
H63790.03EA 22.3 EUR
H63790.03 H63790.06 H63790.14
2-Methyl-6-chinolincarbonsäure 97%
2-Methylquinoline-6-carboxylic acid

Purity: 97% CAS No.: 635-80-3 Molecular Formula: C11H9NO2 Formula Weight: 187.19 Physical Form: Solid MDL No.: MFCD00014566

Formel: C₁₁H₉NO₂
Molecular Weight: 187,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00748504
CAS-Nummer: 635-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR