H-Gln-OtBu·HCl 98%

Lieferant: Thermo Scientific

Synonyme: L-Glutamine tert-butyl ester hydrochloride, H-Gln-OtBu.HCl, H-Gln-OtBu·HCl

H63868.03 H63868.06
H63868.03EA 132 EUR
H63868.03 H63868.06
H-Gln-OtBu·HCl 98%
H-Gln-OtBu·HCl

Purity: 98% CAS No.: 39741-62-3 Molecular Formula: C9H18N2O3.HCl Formula Weight: 238.71 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00039081

Formel: C₉H₁₉ClN₂O₃
Molecular Weight: 238,712 g/mol
MDL: MFCD00039081
CAS-Nummer: 39741-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR