3-Chlor-6-iodpyridazin 97%

Lieferant: Thermo Scientific
Achtung

H63879.03 H63879.06
H63879.03EA 57.8 EUR
H63879.03 H63879.06
3-Chlor-6-iodpyridazin 97%
3-Chloro-6-iodopyridazine

Purity: 97% CAS No.: 135034-10-5 Molecular Formula: C4H2ClIN2 Formula Weight: 240.43 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 110-120° MDL No.: MFCD08275187

Formel: C₄H₂ClIN₂
Molecular Weight: 240,43 g/mol
Schmelzpunkt: 110…120 °C
MDL: MFCD08275187
CAS-Nummer: 135034-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR