N-Methyl-3-(trifluoromethyl)benzylamine 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H63888.03
H63888.06
H63888.03EA
90.3
EUR
H63888.03
H63888.06
N-Methyl-3-(trifluoromethyl)benzylamine 95%
N-Methyl-3-(trifluoromethyl)benzylamine
Purity: 95%
CAS No.: 90390-07-1
Molecular Formula: C9H10F3N
Formula Weight: 189.18
Physical Form: Oil
Appearance: Colourless
UN No.: UN2735
MDL No.: MFCD04115410
Formel:
C₉H₁₀F₃N Molecular Weight: 189.181 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115410 CAS-Nummer: 90390-07-1 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...