4-(4-Fluorophenyl)piperidin-4-ol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(4-Fluorophenyl)-4-hydroxypiperidine

H63892.MD H63892.03 H63892.06
H63892.MDEA 35.3 EUR
H63892.MD H63892.03 H63892.06
4-(4-Fluorophenyl)piperidin-4-ol 97%
4-(4-Fluorophenyl)piperidin-4-ol

Purity: 97% CAS No.: 3888-65-1 Molecular Formula: C11H14FNO Formula Weight: 195.23 Physical Form: Solid MDL No.: MFCD00023146

Formel: C₁₁H₁₄FNO
Molecular Weight: 195.237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023146
CAS-Nummer: 3888-65-1
EINECS: 223-431-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR