Fmoc-N-Me-D-Leu-OH 97%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Fmoc-N-methyl-D-leucine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methylpentanoic acid, Fmoc-D-N-Me-Leu-OH

H63895.03 H63895.06 H63895.14
H63895.03EA 125 EUR
H63895.03 H63895.06 H63895.14
Fmoc-N-Me-D-Leu-OH 97%
Fmoc-N-Me-D-Leu-OH

Purity: 97% CAS No.: 103478-63-3 Molecular Formula: C22H25NO4 Formula Weight: 367.44 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Off-white Beilstein Reference: 7350417 MDL No.: MFCD00235877

Formel: C₂₂H₂₅NO₄
Molecular Weight: 367.445 g/mol
MDL: MFCD00235877
CAS-Nummer: 103478-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR