(±)-β-Aminophenethylalkohol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

H63932.06 H63932.14
H63932.06EA 37 EUR
H63932.06 H63932.14
(±)-β-Aminophenethylalkohol 95%
(±)-β-Aminophenethylalkohol

Purity: 95% CAS No.: 7568-92-5 Molecular Formula: C8H11NO Formula Weight: 137.18 Physical Form: Solid Appearance: Off-white Melting Point: 49-53° Beilstein Reference: 971219 MDL No.: MFCD00130145

Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Schmelzpunkt: 49…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00130145
CAS-Nummer: 7568-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR