1H-Indol-1-ylessigsäure 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Indole-1-acetic Acid, 2-(1H-Indol-1-yl)acetic acid
H63935.MD
H63935.03
H63935.MDEA
114
EUR
H63935.MD
H63935.03
1H-Indol-1-ylessigsäure 95%
(1-Indolyl)acetic acid
Purity: 95%
CAS No.: 24297-59-4
Molecular Formula: C10H9NO2
Formula Weight: 175.18
Physical Form: Solid
Beilstein Reference: 142924
MDL No.: MFCD00047262
Formel:
C₁₀H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047262 CAS-Nummer: 24297-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...