3-(3-Pyridyl)-L-alanine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-2-Amino-3-(pyridin-3-yl)propanoic acid, H-β-(3-Pyridyl)-Ala-OH, H-3-Pal-OH

H63941.MD H63941.03 H63941.06
H63941.MDEA 61.8 EUR
H63941.MD H63941.03 H63941.06
3-(3-Pyridyl)-L-alanine 98%
3-(3-Pyridyl)-L-alanine

Purity: 98% CAS No.: 64090-98-8 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Physical Form: Powder Appearance: Off-white Melting Point: 248-252° dec. Beilstein Reference: 4351583 MDL No.: MFCD00079676

Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 248…252 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00079676
CAS-Nummer: 64090-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR