1H-Indazol-3-carbonsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Indazole-3-carboxylic acid
H64001.06
H64001.14
H64001.22
H64001.06EA
36.8
EUR
H64001.06
H64001.14
H64001.22
1H-Indazol-3-carbonsäure 98%
1H-Indazole-3-carboxylic acid
Purity: 98%
CAS No.: 4498-67-3
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Physical Form: Powder
Appearance: Pale brown
Melting Point: 266-270° lit.
Beilstein Reference: 6354
MDL No.: MFCD00211066
Formel:
C₈H₆N₂O₂ Molecular Weight: 162.15 g/mol Schmelzpunkt: 266…270 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00211066 CAS-Nummer: 4498-67-3 EINECS: 224-794-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...