Thiazole-4-carboxaldehyde 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Thiazole-4-carbaldehyde

H64028.03 H64028.06
H64028.03EA 96.9 EUR
H64028.03 H64028.06
Thiazole-4-carboxaldehyde 95%
Thiazole-4-carboxaldehyde

Purity: 95% CAS No.: 3364-80-5 Molecular Formula: C4H3NOS Formula Weight: 113.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Pale brown Melting Point: 61-65° Beilstein Reference: 107137 MDL No.: MFCD00626896

Formel: C₄H₃NOS
Molecular Weight: 113,13 g/mol
Schmelzpunkt: 61…65 °C
MDL: MFCD00626896
CAS-Nummer: 3364-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR