4-Brom-2,3-difluorbenzaldehyd 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64044.03
H64044.06
H64044.03EA
64.3
EUR
H64044.03
H64044.06
4-Brom-2,3-difluorbenzaldehyd 98%
4-Bromo-2,3-difluorobenzaldehyde
Purity: 98%
CAS No.: 644985-24-0
Molecular Formula: C7H3BrF2O
Formula Weight: 221.00
Physical Form: Powder
Appearance: Off-white
Melting Point: 66-67°
Beilstein Reference: 9551272
MDL No.: MFCD09258953
Formel:
C₇H₃BrF₂O Molecular Weight: 221.00 g/mol Schmelzpunkt: 66…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258953 CAS-Nummer: 644985-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...