1-Brom-2-fluor-5-nitrobenzol 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Brom-1-fluor-4-nitrobenzol, 3-Brom-4-fluornitrobenzol
H64045.14
H64045.22
H64045.14EA
54.4
EUR
H64045.14
H64045.22
1-Brom-2-fluor-5-nitrobenzol 95%
1-Brom-2-fluor-5-nitrobenzol
Purity: 95%
CAS No.: 701-45-1
Molecular Formula: C6H3BrFNO2
Formula Weight: 220.00
Physical Form: Solid
Beilstein Reference: 2089367
MDL No.: MFCD00055433
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055433 CAS-Nummer: 701-45-1 EINECS: 211-855-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...