2-Chlor-6-methoxy-3-nitropyridin 98%
Lieferant: Thermo Fisher Scientific
Achtung
H64076.14 H64076.22
H64076.14EA 78.1 EUR
H64076.14 H64076.22
2-Chlor-6-methoxy-3-nitropyridin 98%
2-Chloro-6-methoxy-3-nitropyridine
Purity: 98% CAS No.: 38533-61-8 Molecular Formula: C6H5ClN2O3 Formula Weight: 188.57 Physical Form: Powder Appearance: Off-white to yellow Melting Point: 78-80° lit. Beilstein Reference: 1454907 UN No.: UN2811 MDL No.: MFCD00130268
Formel: C₆H₅ClN₂O₃
Molecular Weight: 188.57 g/mol
Schmelzpunkt: 78…80 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00130268
CAS-Nummer: 38533-61-8
EINECS: 253-989-8
ADR: 6.1 III