Methyl-4,6-dichlornicotinat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 4,6-dichloronicotinate
H64077.06
H64077.14
H64077.06EA
104
EUR
H64077.06
H64077.14
Methyl-4,6-dichlornicotinat 95%
Methyl-4,6-dichlornicotinat
Purity: 95%
CAS No.: 65973-52-6
Molecular Formula: C7H5Cl2NO2
Formula Weight: 206.02
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 13279782
MDL No.: MFCD04125732
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206,03 g/mol |
MDL:
MFCD04125732 CAS-Nummer: 65973-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...